Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421069.1 | XP_022276777.1 | 2 - 3 | 432 | 80 | |
| XM_022421070.1 | XP_022276778.1 | 2 - 3 | 432 | 80 | ||
| XM_547466.5 | XP_547466.3 | 2 - 3 | 433 | 80 | ||
More Information
Set Membership: |
|

FMO4
Genomic Map