Assay Details
| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639754.3 | XP_005639811.1 | 3 - 4 | 530 | 73 | |
| XM_014110427.2 | XP_013965902.1 | 3 - 4 | 411 | 73 | ||
| XM_014110428.2 | XP_013965903.1 | 3 - 4 | 525 | 73 | ||
| XM_843562.5 | XP_848655.2 | 3 - 4 | 516 | 73 | ||
More Information
Set Membership: |
|

DCUN1D1
Genomic Map