Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621590.2 | XP_005621647.1 | 2 - 3 | 632 | 55 | |
| XM_547143.5 | XP_547143.3 | 2 - 3 | 632 | 55 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map