Assay Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620537.2 | XP_005620594.1 | 466 | 221 | ||
| XM_022419070.1 | XP_022274778.1 | 479 | 221 | |||
| XM_546773.6 | XP_546773.2 | 592 | 221 | |||
More Information
Set Membership: |
|

ZNF276
Genomic Map