Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624428.3 | XP_005624485.2 | 433 | 77 | ||
| XM_014116537.2 | XP_013972012.1 | 772 | 77 | |||
| XM_014116538.2 | XP_013972013.1 | 772 | 77 | |||
| XM_022422953.1 | XP_022278661.1 | 751 | 77 | |||
| XM_022422954.1 | XP_022278662.1 | 735 | 77 | |||
| XM_022422955.1 | XP_022278663.1 | 580 | 77 | |||
| XM_022422956.1 | XP_022278664.1 | 772 | 77 | |||
More Information
Set Membership: |
|

TMEM106A
Genomic Map