Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421917.1 | XP_022277625.1 | 2 - 3 | 787 | 160 | |
| XM_547771.5 | XP_547771.2 | 2 - 3 | 396 | 160 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CFL2
Genomic Map