Assay Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119462.2 | XP_013974937.1 | 4 - 5 | 1500 | 86 | |
| XM_022427740.1 | XP_022283448.1 | 4 - 5 | 1496 | 86 | ||
More Information
Set Membership: |
|

SLFNL1
Genomic Map