Assay Details
| Gene Symbol: PDILT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621484.2 | XP_005621541.1 | 10 - 11 | 1516 | 79 | |
| XM_005621485.2 | XP_005621542.1 | 9 - 10 | 1493 | 79 | ||
| XM_014114588.2 | XP_013970063.1 | 9 - 10 | 1340 | 79 | ||
| XM_022420027.1 | XP_022275735.1 | 9 - 10 | 1521 | 79 | ||
| XM_547107.5 | XP_547107.1 | 9 - 10 | 1442 | 79 | ||
More Information
Set Membership: |
|

PDILT
Genomic Map