Assay Details
Gene Symbol: MAP7D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111728.2 | XP_013967203.1 | 12 - 13 | 1738 | 115 | |
XM_014111732.2 | XP_013967207.1 | 10 - 11 | 1494 | 115 | ||
XM_014111735.2 | XP_013967210.1 | 11 - 12 | 1591 | 115 | ||
XM_014111736.2 | XP_013967211.1 | 9 - 10 | 1371 | 115 | ||
XM_014111737.2 | XP_013967212.1 | 9 - 10 | 1347 | 115 | ||
XM_014111738.2 | XP_013967213.1 | 9 - 10 | 1381 | 115 | ||
XM_022415793.1 | XP_022271501.1 | 12 - 13 | 1810 | 115 | ||
XM_022415795.1 | XP_022271503.1 | 12 - 13 | 1786 | 115 | ||
XM_022415796.1 | XP_022271504.1 | 12 - 13 | 1777 | 115 | ||
XM_022415797.1 | XP_022271505.1 | 11 - 12 | 1711 | 115 | ||
XM_022415798.1 | XP_022271506.1 | 11 - 12 | 1687 | 115 | ||
XM_022415799.1 | XP_022271507.1 | 11 - 12 | 1687 | 115 | ||
XM_022415800.1 | XP_022271508.1 | 11 - 12 | 1663 | 115 | ||
XM_022415801.1 | XP_022271509.1 | 10 - 11 | 1600 | 115 | ||
XM_022415802.1 | XP_022271510.1 | 10 - 11 | 1588 | 115 | ||
XM_022415803.1 | XP_022271511.1 | 10 - 11 | 1576 | 115 | ||
XM_022415804.1 | XP_022271512.1 | 10 - 11 | 1564 | 115 | ||
XM_022415805.1 | XP_022271513.1 | 9 - 10 | 1453 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
