Assay Details
| Gene Symbol: FAM3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639770.4 | XP_003639818.1 | 7 - 8 | 700 | 68 | |
| XM_005632375.3 | XP_005632432.1 | 7 - 8 | 703 | 68 | ||
| XM_022406508.1 | XP_022262216.1 | 8 - 9 | 787 | 68 | ||
| XM_022406509.1 | XP_022262217.1 | 8 - 9 | 623 | 68 | ||
| XM_022406510.1 | XP_022262218.1 | 9 - 10 | 651 | 68 | ||
| XM_022406511.1 | XP_022262219.1 | 8 - 9 | 571 | 68 | ||
More Information
Set Membership: |
|

FAM3D
Genomic Map