Assay Details
| Gene Symbol: ELAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416907.1 | XP_022272615.1 | 534 | 73 | ||
| XM_022416909.1 | XP_022272617.1 | 165 | 73 | |||
| XM_022416912.1 | XP_022272620.1 | 317 | 73 | |||
| XM_022416914.1 | XP_022272622.1 | 421 | 73 | |||
| XM_022416919.1 | XP_022272627.1 | 255 | 73 | |||
| XM_022416925.1 | XP_022272633.1 | 250 | 73 | |||
More Information
Set Membership: |
|

ELAC1
Genomic Map