Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633902.3 | XP_005633959.1 | 761 | 60 | ||
| XM_014106378.2 | XP_013961853.1 | 1731 | 60 | |||
| XM_014106379.2 | XP_013961854.1 | 1690 | 60 | |||
| XM_022408311.1 | XP_022264019.1 | 1560 | 60 | |||
| XM_022408312.1 | XP_022264020.1 | 1706 | 60 | |||
| XM_022408313.1 | XP_022264021.1 | 1534 | 60 | |||
| XM_022408314.1 | XP_022264022.1 | 575 | 60 | |||
| XM_846288.5 | XP_851381.2 | 781 | 60 | |||
More Information
Set Membership: |
|

SLC25A30
Genomic Map