Assay Details
| Gene Symbol: TMBIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427340.1 | XP_022283048.1 | 7 - 8 | 975 | 130 | |
| XM_022427341.1 | XP_022283049.1 | 7 - 8 | 962 | 130 | ||
| XM_847212.5 | XP_852305.2 | 7 - 8 | 830 | 130 | ||
More Information
Set Membership: |
|

TMBIM7
Genomic Map