Assay Details
| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623974.3 | XP_005624031.1 | 1697 | 87 | ||
| XM_014116222.2 | XP_013971697.1 | 1782 | 87 | |||
| XM_014116223.2 | XP_013971698.1 | 1782 | 87 | |||
| XM_014116224.2 | XP_013971699.2 | 1511 | 87 | |||
| XM_014116225.2 | XP_013971700.1 | 1782 | 87 | |||
| XM_022422611.1 | XP_022278319.1 | 1782 | 87 | |||
| XM_022422613.1 | XP_022278321.1 | 1782 | 87 | |||
More Information
Set Membership: |
|

CCDC57
Genomic Map