Assay Details
| Gene Symbol: SMYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630465.3 | XP_005630522.1 | 6 - 7 | 1032 | 73 | |
| XM_847011.5 | XP_852104.1 | 7 - 8 | 1070 | 73 | ||
More Information
Set Membership: |
|

SMYD1
Genomic Map