Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423687.1 | XP_022279395.1 | 1 - 2 | 250 | 71 | |
| XM_022423688.1 | XP_022279396.1 | 1 - 2 | 251 | 71 | ||
| XM_844733.5 | XP_849826.1 | 1 - 2 | 249 | 71 | ||
More Information
Set Membership: |
|

MED27
Genomic Map