Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433337.4 | XP_003433385.1 | 2 - 3 | 321 | 85 | |
| XM_005635702.3 | XP_005635759.1 | 2 - 3 | 240 | 85 | ||
| XM_005635703.3 | XP_005635760.1 | 2 - 3 | 240 | 85 | ||
| XM_005635704.3 | XP_005635761.1 | 2 - 3 | 246 | 85 | ||
| XM_022409870.1 | XP_022265578.1 | 1 - 2 | 136 | 85 | ||
| XM_022409871.1 | XP_022265579.1 | 1 - 2 | 136 | 85 | ||
| XM_022409872.1 | XP_022265580.1 | 2 - 3 | 229 | 85 | ||
| XM_022409873.1 | XP_022265581.1 | 3 - 4 | 376 | 85 | ||
| XM_846111.4 | XP_851204.2 | 2 - 3 | 264 | 85 | ||
More Information
Set Membership: |
|

PEBP4
Genomic Map