Assay Details
| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_847063.5 | XP_852156.1 | 1 - 2 | 353 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISAL2A| Gene Symbol: SHISAL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_847063.5 | XP_852156.1 | 1 - 2 | 353 | 61 | |
Set Membership: |
|