Assay Details
| Gene Symbol: C27H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636898.3 | XP_005636955.1 | 3 - 4 | 283 | 101 | |
| XM_846455.5 | XP_851548.3 | 3 - 4 | 520 | 101 | ||
More Information
Set Membership: |
|

C27H12orf54
Genomic Map