Assay Details
| Gene Symbol: C27H12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636898.3 | XP_005636955.1 | 8 - 9 | 604 | 102 | |
| XM_022411267.1 | XP_022266975.1 | 7 - 8 | 807 | 102 | ||
| XM_846455.5 | XP_851548.3 | 8 - 9 | 841 | 102 | ||
More Information
Set Membership: |
|

C27H12orf54
Genomic Map