Assay Details
| Gene Symbol: MRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116961.2 | XP_013972436.1 | 1 - 2 | 442 | 68 | |
| XM_845298.4 | XP_850391.1 | 3 - 4 | 379 | 68 | ||
More Information
Set Membership: |
|

MRPS2
Genomic Map