Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411277.1 | XP_022266985.1 | 3 - 4 | 679 | 69 | |
| XM_022411278.1 | XP_022266986.1 | 3 - 4 | 679 | 69 | ||
| XM_849704.5 | XP_854797.1 | 3 - 4 | 679 | 69 | ||
More Information
Set Membership: |
|

ADA2
Genomic Map