Assay Details
| Gene Symbol: CABLES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409045.1 | XP_022264753.1 | 9 - 10 | 1467 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CABLES2| Gene Symbol: CABLES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409045.1 | XP_022264753.1 | 9 - 10 | 1467 | 63 | |
Set Membership: |
|