Assay Details
| Gene Symbol: GEMIN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638826.4 | XP_003638874.1 | 2 - 3 | 222 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003638826.4 transcripts.
Set Membership: |
|

GEMIN7
Genomic Map