Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406927.1 | XP_022262635.1 | 3 - 4 | 365 | 75 | |
| XM_847645.4 | XP_852738.1 | 3 - 4 | 552 | 75 | ||
More Information
Set Membership: |
|

TMEM38A
Genomic Map