Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_848001.4 | XP_853094.3 | 7 - 8 | 1255 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP27A1| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_848001.4 | XP_853094.3 | 7 - 8 | 1255 | 82 | |
Set Membership: |
|