Assay Details
| Gene Symbol: LOC611363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420286.1 | XP_022275994.1 | 6 - 7 | 766 | 59 | |
| XM_022420287.1 | XP_022275995.1 | 6 - 7 | 706 | 59 | ||
| XM_022420288.1 | XP_022275996.1 | 6 - 7 | 697 | 59 | ||
| XM_022420289.1 | XP_022275997.1 | 6 - 7 | 679 | 59 | ||
| XM_022420290.1 | XP_022275998.1 | 6 - 7 | 908 | 59 | ||
| XM_022420291.1 | XP_022275999.1 | 6 - 7 | 803 | 59 | ||
| XM_022420292.1 | XP_022276000.1 | 6 - 7 | 665 | 59 | ||
More Information
Set Membership: |
|

LOC611363
Genomic Map