Assay Details
| Gene Symbol: TSKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616294.2 | XP_005616351.1 | 7 - 8 | 1302 | 103 | |
| XM_005616295.2 | XP_005616352.1 | 7 - 8 | 1229 | 103 | ||
| XM_005616296.2 | XP_005616353.1 | 7 - 8 | 1301 | 103 | ||
| XM_005616297.2 | XP_005616354.1 | 7 - 8 | 1300 | 103 | ||
| XM_005616298.3 | XP_005616355.1 | 7 - 8 | 1301 | 103 | ||
| XM_005616299.2 | XP_005616356.1 | 7 - 8 | 1299 | 103 | ||
| XM_005616300.2 | XP_005616357.1 | 7 - 8 | 1301 | 103 | ||
| XM_005616301.2 | XP_005616358.1 | 7 - 8 | 1226 | 103 | ||
| XM_849620.4 | XP_854713.1 | 7 - 8 | 1226 | 103 | ||
More Information
Set Membership: |
|

TSKS
Genomic Map