Assay Details
| Gene Symbol: METTL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426448.1 | XP_022282156.1 | 7 - 8 | 967 | 67 | |
| XM_022426449.1 | XP_022282157.1 | 2 - 3 | 325 | 67 | ||
More Information
Set Membership: |
|

METTL24
Genomic Map