Assay Details
| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616674.3 | XP_005616731.1 | 1 - 2 | 161 | 65 | |
| XM_022404444.1 | XP_022260152.1 | 1 - 2 | 161 | 65 | ||
| XM_850347.5 | XP_855440.1 | 1 - 2 | 161 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005616674.3, XM_022404444.1, XM_850347.5 transcripts.
Set Membership: |
|

COX7A1
Genomic Map