Assay Details
| Gene Symbol: ALDH1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286977.1 | NP_001273906.1 | 8 - 9 | 897 | 62 | |
| GenBank mRNA | DN864805.1 | - | 2 - 3 | 134 | 62 | |
More Information
Set Membership: |
|

ALDH1A1
Genomic Map