Assay Details
| Gene Symbol: UBQLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_853048.5 | XP_858141.2 | 9 - 10 | 1447 | 92 | |
| XM_853126.5 | XP_858219.1 | 8 - 9 | 1363 | 92 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UBQLN1
Genomic Map