Assay Details
| Gene Symbol: SOCS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122454.2 | XP_013977929.1 | 1769 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014122454.2 transcripts.
Set Membership: |
|

SOCS6
Genomic Map