Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635437.3 | XP_005635494.1 | 252 | 67 | ||
| XM_022409609.1 | XP_022265317.1 | 482 | 67 | |||
| XM_022409610.1 | XP_022265318.1 | 531 | 67 | |||
| XM_534493.6 | XP_534493.2 | 312 | 67 | |||
More Information
Set Membership: |
|

ALG5
Genomic Map