Assay Details
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629469.3 | XP_005629526.1 | 675 | 65 | ||
| XM_005629470.3 | XP_005629527.1 | 675 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629469.3 | XP_005629526.1 | 675 | 65 | ||
| XM_005629470.3 | XP_005629527.1 | 675 | 65 | |||
Set Membership: |
|