Assay Details
| Gene Symbol: ACTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618839.2 | XP_005618896.1 | 156 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005618839.2 transcripts.
Set Membership: |
|

ACTA1
Genomic Map