Assay Details
| Gene Symbol: NFE2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640352.3 | XP_005640409.1 | 2 - 3 | 414 | 75 | |
| XM_014110726.1 | XP_013966201.1 | 2 - 3 | 522 | 75 | ||
| XM_022414833.1 | XP_022270541.1 | 2 - 3 | 391 | 75 | ||
More Information
Set Membership: |
|

NFE2L2
Genomic Map