Assay Details
| Gene Symbol: UBFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621409.3 | XP_005621466.1 | 3 - 4 | 798 | 101 | |
| XM_005621410.3 | XP_005621467.1 | 3 - 4 | 798 | 101 | ||
| XM_005621411.3 | XP_005621468.1 | 3 - 4 | 798 | 101 | ||
| XM_005621412.3 | XP_005621469.1 | 3 - 4 | 798 | 101 | ||
| XM_005621413.3 | XP_005621470.1 | 3 - 4 | 798 | 101 | ||
| XM_005621414.3 | XP_005621471.1 | 3 - 4 | 798 | 101 | ||
| XM_022419948.1 | XP_022275656.1 | 3 - 4 | 798 | 101 | ||
| XM_022419949.1 | XP_022275657.1 | 3 - 4 | 798 | 101 | ||
| XM_856141.6 | XP_861234.2 | 3 - 4 | 798 | 101 | ||
More Information
Set Membership: |
|

UBFD1
Genomic Map