Assay Details
| Gene Symbol: UQCRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119566.2 | XP_013975041.1 | 2 - 3 | 156 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014119566.2 transcripts.
Set Membership: |
|

UQCRH
Genomic Map