Assay Details
| Gene Symbol: SMARCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624251.3 | XP_005624308.1 | 508 | 99 | ||
| XM_005624252.3 | XP_005624309.1 | 364 | 99 | |||
| XM_022422831.1 | XP_022278539.1 | 479 | 99 | |||
| XM_845276.5 | XP_850369.1 | 865 | 99 | |||
More Information
Set Membership: |
|

SMARCD2
Genomic Map