Assay Details
| Gene Symbol: ATP6V1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637384.3 | XP_005637441.1 | 6 - 7 | 572 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V1E1| Gene Symbol: ATP6V1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637384.3 | XP_005637441.1 | 6 - 7 | 572 | 143 | |
Set Membership: |
|