Assay Details
| Gene Symbol: IL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003157.2 | NP_001003157.2 | 6 - 7 | 690 | 96 | |
| XM_005630066.3 | XP_005630123.1 | 5 - 6 | 526 | 96 | ||
| GenBank mRNA | AF047011.1 | - | 5 - 6 | 602 | 96 | |
| DQ923806.1 | - | 6 - 7 | 690 | 96 | ||
| EF068230.1 | - | 5 - 6 | 516 | 96 | ||
More Information
Set Membership: |
|

IL1A
Genomic Map