Assay Details
| Gene Symbol: MSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408661.1 | XP_022264369.1 | 3 | 1770 | 72 | |
| XM_022408662.1 | XP_022264370.1 | 2 | 1415 | 72 | ||
| XM_542793.6 | XP_542793.3 | 2 | 2144 | 72 | ||
More Information
Set Membership: |
|

MSL2
Genomic Map