Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626755.3 | XP_005626812.1 | 5 - 6 | 1487 | 86 | |
| XM_005626756.2 | XP_005626813.1 | 5 - 6 | 1540 | 86 | ||
| XM_005626757.2 | XP_005626814.1 | 5 - 6 | 1366 | 86 | ||
| XM_005626758.2 | XP_005626815.1 | 5 - 6 | 1333 | 86 | ||
| XM_847692.5 | XP_852785.1 | 5 - 6 | 1421 | 86 | ||
More Information
Set Membership: |
|

FAM214B
Genomic Map