Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639071.4 | XP_003639119.1 | 3 | 1324 | 120 | |
| XM_005620815.3 | XP_005620872.1 | 4 | 1503 | 120 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GFOD2
Genomic Map