Assay Details
| Gene Symbol: CCM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639538.4 | XP_003639586.2 | 392 | 140 | ||
| XM_005629571.3 | XP_005629628.1 | 1209 | 140 | |||
| XM_022403750.1 | XP_022259458.1 | 420 | 140 | |||
| XM_022403751.1 | XP_022259459.1 | 395 | 140 | |||
| XM_022403753.1 | XP_022259461.1 | 494 | 140 | |||
| XM_022403754.1 | XP_022259462.1 | 130 | 140 | |||
More Information
Set Membership: |
|

CCM2
Genomic Map