Assay Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627826.2 | XP_005627883.1 | 2146 | 150 | ||
| XM_005627827.3 | XP_005627884.1 | 2081 | 150 | |||
| XM_022426581.1 | XP_022282289.1 | 2147 | 150 | |||
| XM_539103.5 | XP_539103.4 | 2146 | 150 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FBXO43
Genomic Map