Assay Details
| Gene Symbol: ARHGAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623314.3 | XP_005623371.1 | 6 - 7 | 4578 | 145 | |
| XM_845275.5 | XP_850368.1 | 6 - 7 | 4581 | 145 | ||
More Information
Set Membership: |
|

ARHGAP5
Genomic Map