Assay Details
| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411093.1 | XP_022266801.1 | 7 - 8 | 1018 | 137 | |
| XM_535297.6 | XP_535297.2 | 7 - 8 | 852 | 137 | ||
More Information
Set Membership: |
|

OGFOD1
Genomic Map