Assay Details
| Gene Symbol: CALU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427257.1 | XP_022282965.1 | 4 - 5 | 610 | 128 | |
| XM_844242.5 | XP_849335.1 | 4 - 5 | 721 | 128 | ||
| XM_853685.5 | XP_858778.1 | 4 - 5 | 721 | 128 | ||
More Information
Set Membership: |
|

CALU
Genomic Map